Learn More
asecos™ Steel Basin Shelf Levels
The Steel Basin Shelf Levels provide an additional storage level.
Brand:  asecos™ 27876
 View more versions of this product
Product Code. 17806586
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Incl. reinforcing traverses and locking pins
  • Blue-painted steel
Specifications
Specifications
Blue
Basin Shelf
1300.00 mm
1300.00 mm
65.00 mm
600.00 mm
27 kg
Steel
51.18 in.
51.18 in.
2.56 in.
23.62 in.
59.52 lb.
Product Title
asecos™ Steel Basin Shelf Levels > Weight: 27 kg