Learn More
Bochem™ Steel Closing Tong
Manufactured from steel. BOCHEM™ Steel Closing Tong is used with aluminum caps.
Brand:  Bochem™ 12963
Product Code. 10090911
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • With adjust screw
  • For aluminum caps
Specifications
Specifications
20 mm
Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Steel Closing Tong > Diameter: 20mm