Learn More
Fisherbrand™ Stainless Steel Straight Tongs
Get maximum results using laboratory tongs in high temperature chemical reactions. Made of durable stainless steel, these Clifton™ Stainless Steel Straight Tongs ensure precise control of grasping, lifting, turning, and transferring heat-resistant and other materials in a laboratory environment.
57.55 DKK - 87.15 DKK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
11593492
|
Fisherbrand™
3030 |
150 mm |
57.55 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
11503502
|
Fisherbrand™
3031 |
200 mm |
87.15 DKK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Description
- Laboratory straight tongs
- Stainless steel 304 spec 18/8 non-magnetic, bright finish
- Designed for corrosion resistance and durability ideal in laboratory routines
- Laboratory metalware consumables
- Finest in corrosion-resistant 304 stainless steel
- Provides corrosion resistance that prevents product contamination
Specifications
Steel | |
Crucible Tongs | |
1 Ea. |