Learn More
asecos™ Steel Grid Storage Level
The Steel Grid Storage Level provides an additional storage level.
Brand:  asecos™ 27880
 View more versions of this product
Product Code. 17886396
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Incl. reinforcing traverses and locking pins
  • Galvanised steel
Specifications
Specifications
Steel
51.18 in.
51.18 in.
0.79 in.
23.62 in.
26.46 lb.
Storage Level
1300.00 mm
1300.00 mm
20.00 mm
600.00 mm
12 kg
Documents
One moment while we fetch your results.
Product Title
asecos™ Steel Grid Storage Level > Weight: 12 kg