Learn More
Fisherbrand™ Steel Stand Rings with Rod
Provides for ease of connection on ring stands. Fisherbrand™ Steel Stand Rings with Rod are manufactured from plated mild steel.
95.60 DKK - 106.00 DKK
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | ||||||
12388149
|
Fisherbrand™
2010/11 |
100 mm |
95.60 DKK
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
12976836
|
Fisherbrand™
2010/14 |
127 mm |
102.00 DKK
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
12986836
|
Fisherbrand™
2010/17 |
150 mm |
106.00 DKK
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
12956836
|
Fisherbrand™
2010/5 |
70 mm |
95.65 DKK
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
12946836
|
Fisherbrand™
2010/2 |
50 mm |
95.65 DKK
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
||||||
Description
- 8mm diameter stems
- 140mm stem lengths
- Rings in a variety of diameters
Specifications
Steel | |
Stand Ring |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Steel Stand Rings with Rod