Learn More
Usbeck Laborgeraete™ Steel Triangles
24.35 DKK - 29.25 DKK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10303101
|
Usbeck Laborgeraete™
5100 |
40 mm |
24.35 DKK
Each |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
10189740
|
Usbeck Laborgeraete™
5101 |
50 mm |
25.25 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
10423121
|
Usbeck Laborgeraete™
5102 |
60 mm |
26.05 DKK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
10433121
|
Usbeck Laborgeraete™
5104 |
80 mm |
29.25 DKK
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
Specifications
Steel | |
1 Each | |
Triangles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Usbeck Laborgeraete™ Steel Triangles