missing translation for 'onlineSavingsMsg'
Learn More
STEMCELL Technologies STEMdiff™ APEL™2 Medium Product Code.: 17148261

STEMCELL Technologies STEMdiff™ APEL™2 Medium

Product Code. 17148261
500 mL, 500mL
Encompass
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 17148261

Brand: STEMCELL Technologies 05275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Defined, animal component-free medium for differentiation of human ES and iPS cells to multiple lineages

  • STEMdiff™ APEL™ 2 Medium is a fully defined, serum-free and animal component-free medium for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • It is based on the APEL formulation published by Dr
  • Andrew Elefanty and lacks undefined components such as protein-free hybridoma medium
  • STEMdiff™ APEL™ 2 can be used in adherent or embryoid body (EB)-based protocols, such as with AggreWell™
  • It can be used with a variety of different induction factors or cytokines to support differentiation along ectoderm, mesoderm and endoderm lineages

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 500 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
STEMCELL Technologies STEMdiff™ APEL™2 Medium

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.