Learn More
STEMCELL Technologies STEMdiff™ Neural Progenitor Freezing Medium
For cryopreservation of neural progenitor cells generated using STEMdiff™ Neural Induction Medium
Brand:  STEMCELL Technologies 05838
Encompass_Preferred
Product Code. 15385811
  • 3538.35 DKK / Each
This item is not returnable. View return policy
Description
Description
  • STEMdiff™ Neural Progenitor Freezing Medium is a serum-free medium for cryopreservation of neural progenitor cells (NPCs) derived from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • This freezing medium is optimized for the cryopreservation of NPCs generated using STEMdiff™ Neural Induction Medium (Catalog #05835) and cultured in STEMdiff™ Neural Progenitor Medium (Catalog #05833)
  • NPCs can be frozen at any point post-neural induction, with reproducibly high recovery rates
  • Post-thaw, NPCs display healthy morphology, express NPC markers, and retain the potential to expand and differentiate into neurons
Specifications
Specifications
Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells
Cryopreservation
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies STEMdiff™ Neural Progenitor Freezing Medium