missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Sterile Blue Gloves
Description
- Packed in pairs and folded
- Made of blue polyethylene and
- Sterility is ensured by gamma irradiation
- Comply with the highest quality standards
- Can be used in all day-to-day laboratory procedures
Specifications
Specifications
Applications | Laboratory |
Color | Blue |
Disposable | Single-use |
Latex Free | Yes |
Product Type | Sterile Gloves |
Sterility | Sterile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'