Learn More
Thermo Scientific™ Sterilin™ Black and White Microtiter™ Plates
Perform both fluorescence work and bioluminescence studies with 96-well Thermo Scientific™ Sterilin™ Black and White Microtiter™ plates.
Brand: Thermo Scientific™ 611F96BK
984.85 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Perform both fluorescence work and bioluminescence studies with 96-well Thermo Scientific™ Sterilin™ Black and White Microtiter™ plates. Supplied with no lid; if required, the 642000 lid is a suitable fit.
- Black 96-well plates are ideal for fluorescence work where the black pigment prevents «cross-talk« between the wells — the low background fluorescence minimizes light scattering
- White 96-well plates are ideal for bioluminescence studies where the pigment aids reflectivity and sensitivity
- SBS compliant, ensuring suitable fit with all automation
- Manufactured from virgin polystyrene
- Supplied nonsterile, providing a medium binding surface
- Chimney well design to help reduce contamination
Ordering Alerts:
Supplied without a lid; if required, the 642000 lid is a suitable fit.
Specifications
Standard | |
50 Cases, 5 Pk. | |
Microplate | |
400 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
5 |
8 x 12 | |
Black | |
Polystyrene | |
96 | |
Non-treated | |
50 | |
50/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M541720 | Certificate of Compliance | 07/01/2025 | 611F96BK |
M452990 | Certificate of Compliance | 03/12/2024 | 611F96BK |
M510120 | Certificate of Compliance | 03/12/2024 | 611F96BK |
M420620 | Certificate of Compliance | 03/12/2024 | 611F96BK |
M498400 | Certificate of Compliance | 03/12/2024 | 611F96BK |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now