Learn More
Buerkle™ SteriPlast™ Disposable Sample Scoop
Sample powder, granules, and pastes with these sterile polystyrene scoops. Available with or without transparent lids to help ensure a sterile and contamination-free sampling procedure.
182.00 DKK - 769.00 DKK
Product Code | Brand | Capacity (Metric) | Length (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Includes | Price | Quantity & Availability | ||||
15366317
![]() ![]() |
Buerkle™
5378-1001 |
25 mL | 141 mm | Without Lid |
182.00 DKK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17280584
![]() ![]() |
Buerkle™
5378-1020 |
2000 mL |
769.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
15694104
![]() ![]() |
Buerkle™
5378-1019 |
1000 mL | 332 mm | Without Lid |
662.00 DKK
Pack of 10 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
15376317
![]() ![]() |
Buerkle™
5378-1002 |
25 mL | 142 mm | With Lid |
276.00 DKK
Pack of 10 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||
15386317
![]() ![]() |
Buerkle™
5378-1003 |
50 mL | 160 mm | Without Lid |
189.00 DKK
Pack of 10 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||
15396317
![]() ![]() |
Buerkle™
5378-1004 |
50 mL | 161 mm | With Lid |
284.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
15306327
![]() ![]() |
Buerkle™
5378-1005 |
100 mL | 205 mm | Without Lid |
195.00 DKK
Pack of 10 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||
15390937
![]() ![]() |
Buerkle™
5378-1006 |
100 mL | 206 mm | With Lid |
293.00 DKK
Pack of 10 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||
15316327
![]() ![]() |
Buerkle™
5378-1007 |
150 mL | 216 mm | Without Lid |
199.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||
15326327
![]() ![]() |
Buerkle™
5378-1008 |
150 mL | 218 mm | With Lid |
299.00 DKK
Pack of 10 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||
15581721
![]() ![]() |
Buerkle™
5378-1013 |
250 mL | 232 mm | Without Lid |
243.00 DKK
Pack of 10 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||
15591721
![]() ![]() |
Buerkle™
5378-1014 |
250 mL | 253 mm | With Lid |
414.00 DKK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
15501731
![]() ![]() |
Buerkle™
5378-1015 |
500 mL | 280 mm | Without Lid |
430.00 DKK
Pack of 10 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
15511731
![]() ![]() |
Buerkle™
5378-1016 |
500 mL | 283 mm | With Lid |
733.00 DKK
Pack of 10 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||
Description
- Available with or without a transparent cover to help protect samples from contamination
- Ergonomically designed handle with a slightly angled edge
- Scoop stands horizontally and can be laid down without materials trickling out
- Ideal fo ruse with powders, granules, and pastes
- Individually packed for one-way needs
- Sterilized by gamma radiation
- Production, assembling, and packaging according to cleanroom class 7 (10.000)
Specifications
Polystyrene | |
Not Autoclavable | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Buerkle™ SteriPlast™ Disposable Sample Scoop