Learn More
Sampling Systems™ SteriWare™ Counting Triangle
BIOPHARMA
The SteriWare™ Counting Triangle is ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
Brand:  Sampling Systems™ 8066A-1S
Product Code. 15960531
  • 1200.00 DKK / Pack of 30
Only null left
Estimated Shipment: 19-09-2025
Description
  • Suitable for left- and right-handed operators
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming polystyrene
  • Completely sterile
  • Provided 30 per box
Specifications
Specifications
Counting Triangle
Polystyrene
30/box
235 mm side length, 15 mm height
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number118378 Certificate TypeCertificate of Compliance Date07/03/2025 Catalog Number8066A-1S
Lot Number071985 Certificate TypeCertificate of Compliance Date07/12/2022 Catalog Number8066A-1S
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Counting Triangle > Counting triangle