missing translation for 'onlineSavingsMsg'
Learn More

STK36 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1288.30 DKK - 2621.96 DKK

Specifications

Antigen STK36
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18310945
View Documents Promotion Details
Bio-Techne
NBP3-15339-20UL
20 μg
1288.30 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
18309165
View Documents Promotion Details
Bio-Techne
NBP3-15339-100UL
100 μg
2621.96 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STK36 Polyclonal antibody specifically detects STK36 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

STK36
Polyclonal
Purified
RUO
Human, Mouse
EC 2.7.11, EC 2.7.11.1, Fused homolog, FUserine/threonine kinase 36, fused homolog, KIAA1278DKFZp434N0223, serine/threonine kinase 36, serine/threonine kinase 36 (fused homolog, Drosophila), serine/threonine-protein kinase 36
Recombinant fusion protein containing a sequence corresponding to amino acids 250-450 of human STK36 (NP_056505.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Protein Kinase
PBS, 50% glycerol, pH7.3
27148
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications