Learn More
Poulten Graf™ Stopper Caps
Brand:  Poulten Graf™ 101 08918K
 View more versions of this product
Product Code. 10178101
  • 319.00 DKK / Pack of 10
Only null left
Estimated Shipment: 15-08-2025
Specifications
Stopper Cap
10 , 30 mL
1 Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Poulten Graf™ Stopper Caps > Capacity: 10/0mL