Learn More
Fisherbrand™ Tubby™ Storage Containers
Store, organize and transport small items to maintain a tidy lab. Fisherbrand™ Tubby™ Storage Containers feature removable dividers for customized bin compartments. Features tight-fitting lids and heavy-impact, clear polypropylene.
Brand: Fisherbrand™ HS23453
227.30 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
- Stackable and proportionately sized to utilize space efficiently
- Removable dividers create as big or as small a compartment as needed
- Lid snaps in place to secure the contents for safe transport
- Clear sides for at-a-glance access and inventory monitoring
- Ideal for tubes, tips, pipettes, gloves and other small items
- 5-pack includes 5 tubs, 5 lids and 15 dividers
Specifications
Storage Bin | |
Clear | |
5/Pk. | |
200 mm |
Polypropylene | |
Divider, Lid | |
330 mm | |
115 mm |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now