Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Resin
Strep-Tactin™ Sepharose™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences
This item is not returnable.
View return policy
Description
- Strep-Tactin™ Sepharose™ is a 4% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
- Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
- The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
- The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
- To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.
Please note that Strep-Tactin™ Sepharose™ is not compatible with HPLC/FPLC due to the low pressure stability.
Specifications
Strep-Tactin™ Sepharose™ 50% suspension | |
50 mL | |
Pk. Affinity Chromatography Columns for Protein Purification | |
Strep-Tactin Sepharose |
Manual Strep-Tactin-Purification 2-1201-025
File size: 1.4 MB
Short Protocol Strep-Tactin-Purification 2-1201-025
File size: 695 kB
Product Content Correction
Product Title
IBA Lifesciences Strep-Tactin™ Sepharose™ Resin > Quantity: 50 mL
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction