Learn More
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using this test kit.
Brand: Thermo Scientific™ DR0591G
710.55 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.
- Supplied with rapid latex agglutination test
Specifications
Streptococcal Grouping Reagent G | |
Reagents for Streptococcus Testing | |
Group A, B, C, D, F and G | |
50 Tests/Vial |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6192255 | Certificate of Analysis | 23/04/2025 | DR0591G |
6177068 | Certificate of Analysis | 10/02/2025 | DR0591G |
6177067 | Certificate of Analysis | 17/12/2024 | DR0591G |
6143433 | Certificate of Analysis | 04/12/2024 | DR0591G |
6143432 | Certificate of Analysis | 21/10/2024 | DR0591G |