missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ STS 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand: Abnova™
This item is not returnable.
View return policy
Specifications
293T | |
STS 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 64.24kDa | |
Transient overexpression cell lysate was tested with anti-STS antibody (Mfr. No. H00000412-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
NM_000351.3 | |
pCMV-STS full-length | |
100 μL | |
Human |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UL STS 293T Cell Transient Overexpression Lysat
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Mercaptoethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction