Learn More
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators
Quickly and easily replace tube holder without tools using these Tube Holder Discs for Cole-Parmer™ TR-200 Series Tube Rotators.
Brand:  Cole-Parmer™ 00077-21
2161.98 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11446558
  • 2210.00 DKK / Each
Only null left
Estimated Shipment: 15-08-2025
Description
  • Choose the tube holder(s) that accommodates your tube size(s).
Specifications
Specifications
Tube Holder
For 30 Tubes, 26 mm dia.
30
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators > Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators; For Use With: For 30 Tubes, 26 mm dia.