Learn More
Cole-Parmer™ Stuart™ Block Heater Insert
Click to view available options
95 x 75 x 50 mm (Cuvette/Pipette Channels)
Description
- For use with Stuart block heaters
- Manufactured from anodized aluminum
- Separate hole accommodates a thermometer if desired
Specifications
Specifications
Description | Holds Cuvette/Pipette Channels |
Holds | Cuvette/Pipette Channels |
Material | Anodized Aluminum |
No. of Places | Cuvette/Pipette Channels |
Type | Block |
Product Content Correction
Product Title
Cole-Parmer™ Stuart™ Block Heater Insert >
Spot an opportunity for improvement?Share a Content Correction