Learn More
Succimer, MedChemExpress Product Code.: 16478806
Succimer, MedChemExpress
Product Code. 16478806
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16478806
Brand: Medchem Express HYB1768
MedChemExpress Succimer is a widely used chelating agent for the treatment of Pb poisoning.
Succimer is a widely used chelating agent for the treatment of Pb poisoning.
Specifications
Chemical Name or Material Succimer
CAS 304-55-2
Color White
Purity Grade Notes Research
Molecular Formula C4H6O4S2
Quantity 100 mg
Synonym Dimercaptosuccinic acid DMSA
Solubility Information DMSO : ≥ 100 mg/mL (548.79 mM)
SMILES O=C(O)[C@@H](S)[C@@H](S)C(O)=O
Molecular Weight (g/mol) 182.22
Formula Weight 182.22
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Succimer, MedChemExpress