Learn More
Mettler Toledo™ Sulfide Half Cell Electrode
Brand:  Mettler Toledo™ 51107675
9475.42 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11937701
  • 9720.00 DKK / Each
In Stock
Explore more special offers
Specifications
Solid State Membrane
0°C to 80°C
4 to 13
Product Title
Mettler Toledo™ Sulfide Half Cell Electrode > Membrane: Solid State