missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric acid, 0.1N Standardized Solution, Thermo Scientific Chemicals Product Code.: 11373797

Sulfuric acid, 0.1N Standardized Solution, Thermo Scientific Chemicals

Product Code. 11373797
20 L
Click to view available options
Quantity:
1 L
4 L
20 L
This item is not returnable. View return policy

Product Code. 11373797

Brand: Thermo Scientific Alfa Aesar 035651.M4

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sulfuric acid, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Quantity 20 L
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1g/cm3 at 20°C
Physical Form Liquid
Solubility Miscible with water
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 0.05%; Water: 99.95%
Chemical Name or Material Sulfuric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP16L528 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number035651.K2,035651.M4,035651.K7
Lot NumberM31L504 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number035651.K2,035651.M4,035651.K7
Lot Number62400111 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog Number035651.K2,035651.M4,035651.K7
Lot NumberM16L549 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number035651.K2,035651.M4,035651.K7
Lot NumberZ27K511 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number035651.K2,035651.M4,035651.K7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric acid, 0.1N Standardized Solution, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.