missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid, , 0.25M, Chem Lab™ Product Code.: 11953094
compliance-icons

Sulfuric Acid, , 0.25M, Chem Lab™

Product Code. 11953094
1 L, 1 L
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11953094

Brand: Chem Lab CL05.2614.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Appearance Colorless
pH <1
Composition Sulfuric acid 0.25 mol/L
Concentration 24.52g H2SO4/l H2O = 0.5 N (±0.001/20°C)
Density 1.02g/mL
Odor Odorless
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons
Product Identifier
  • Sulphuric acid 0.25 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid, , 0.25M, Chem Lab™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.