Learn More
Sulfuric Acid, 10%, Chem Lab™ Product Code.: 11945059
compliance-icons
Sulfuric Acid, 10%, Chem Lab™
Product Code. 11945059
1 L, 1 L
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11945059
Brand: Chem Lab CL02.2612.1000
Specifications
Appearance Colorless
pH <1
Composition Sulfuric acid 10 vol. % solution
Concentration 185g H2SO4/l H2O
Density 1.11g/mL
Odor Odorless
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons
Product Identifier
  • Sulfuric acid 10 vol. % solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Product Title
Sulfuric Acid, 10%, Chem Lab™ >