Learn More
Diversey™ Suma Alcohol Wipes
No-rinse alcoholic disinfectant wipes
Brand: Diversey™ 100878520
Description
Suma Alcohol Wipes are ready-to-use wipes for disinfection of all hard surfaces like work areas, small equipment, utensils, and handholds.
Key properties
- Suma Alcohol Wipes are perfect for disinfection of all food contact areas and can be used as complement to your cleaning and disinfection procedures.
- The product is based on ethanol to provide fast and effective disinfection.
- It is suitable for the disinfection of alcohol-safe, food contact surfaces and articles such as chopping boards, utensils, thermometers, tables and other smaller kitchen equipment.
- It is effective against a wide spectrum of micro-organisms.
Benefits
- Easy to use
- No spraying, safe to use
- Effective against a wide range of micro-organisms
- Quick drying minimizes disruption to work flow
Specifications
Disinfectant Wipe | |
180 mm | |
12 x 150 Wipes |
135 mm | |
135 x 180 mm | |
12 x 150 Wipes for DK, SE, FIN, NO only |
Safety and Handling
- Suma Alcohol Wipes
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : ethanol, propan-2-ol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.