Learn More
uvex Super Fit Safety Spectacles
Shop All Uvex Products
Click to view available options
Frame Color:
White, Blue
White, Lime
Lens Tint:
Clear
Unit Size:
Each
Description
- Flexible and adjustable nose bridge
- UV protection
- Anti-fog
- Scratch-resistant
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Glasses |
Frame Color | White, Lime |
Model | Superfit 9178-315 |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Frame Material | Plastic |
Show More |
Product Content Correction
Product Title
uvex Super Fit Safety Spectacles >
Spot an opportunity for improvement?Share a Content Correction