Learn More
Velp Scientifica™ Support Rod
For Use With: Velp Heating Magnetic Stirrer and Hot Plate
Brand:  Velp Scientifica™ A00001069
186.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11747303
  • 192.00 DKK / Each
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Only null left
Estimated Shipment: 19-08-2025
Description
For Use With: Velp Heating Magnetic Stirrer and Hot Plate
  • For use with AREX™ CerAlTop™ Hot Plate Stirrers
  • Helps keep round-bottom flasks uprigth during stirring/heating
Specifications
Specifications
12 mm
Velp Heating Magnetic Stirrer and Hot Plate
470 mm
Rod
Documents
Brochures
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Velp Scientifica™ Support Rod > Length: 470 mm