Documents
Scientific Resources
Posters
- 1
- 2
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System.
Brand: Thermo Scientific™ PT0500
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System. The system combines a real-time PCR instrument with bespoke software and convenient pre-filled reagent components that, together with a single PCR protocol for a range of common targets and sample types, streamline the test workflow.
The SureTect System is a unique solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
±2°C | |
10 logs | |
310 mm | |
230 mm | |
SureTect Real Time PCR Assays | |
>5°C/s | |
30 to 110°C | |
5 LEDs | |
Absolute quantification; Relative quantification; Melt curve analysis; Allelic discrimination; High Resolution Melt (HRM) | |
10 to 50ÎĽL | |
10 kg |
1 Copy (Theoretical) | |
PikoReal 24 | |
300 mm | |
4° to 99.9°C | |
PikoReal 24 (24-well Piko PCR Plate) | |
<10 s. | |
4.5deg;C/s | |
520 to 740 nm | |
Automatic Temperature and Pressure Setting | |
Up to 4 Targets | |
24-well |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.