Learn More
Swann-Morton™ Stainless Steel Scalpel Handles, Graduated
Use Swann Morton™ Stainless Steel Scalpel Handles, Graduated to assist with training, marking of the correct surgical site and trauma injury investigations.
Brand:  Swann-Morton™ 0934
 View more versions of this product
Product Code. 11890862
  • 773.00 DKK / Pack of 10
Only null left
Estimated Shipment: 18-08-2025
Description
  • Non-sterile
  • Individually packed
  • Graduated
  • Stainless Steel
Specifications
Specifications
Stainless Steel
# 4
Model No. 4GS /S
Yes
Surgical Scalpel Handle
130 mm
Autoclavable
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Swann-Morton™ Stainless Steel Scalpel Handles, Graduated > No. 4; Autoclavable