Learn More
BICASA Suspended Undercounter Cabinet with Drawers for Kompatto Workstation
Click to view available options
Frame Color:
Anthracite
Blue
Red
Unit Size:
Each
Description
- For Kompatto Series Workstation
- Suspended under the countertop
- Drawers: EN 14749 certified sliding guide - Soft-Close damping system - Single loading capacity of Kg 40,0
- Central locking system as standard
- Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
- Fire-retardant particleboard body
- Light Gray color (2 decorative) - mm 19 thickness
- Frontal drawer panels and doors with ABS rounded rim
Specifications
Specifications
Product Type | Suspended Undercounter Cabinet |
Color | Light Gray |
Frame Color | Red |
Material | Particleboard |
Includes | 3 Soft-close Drawers with Central Lock |
For Use With (Equipment) | Kompatto Series Workstation |
Dimensions (L x W x H) | 600 x (500 + 20) x 530 mm |
Width (English) | 19.7 in. |
Width (Metric) | 500 mm |
Product Content Correction
Product Title
BICASA Suspended Undercounter Cabinet with Drawers for Kompatto Workstation >
Spot an opportunity for improvement?Share a Content Correction