Learn More
BICASA Suspended Undercounter Cabinet with Hinged Door for Kompatto Workstation Product Code.: 30057587
Estimated Shipment: 15-08-2025
Only null left
Add to basket
BICASA Suspended Undercounter Cabinet with Hinged Door for Kompatto Workstation
Product Code. 30057587
Right Opening Door, Blue, Each
Click to view available options
Includes:
Left Opening Door
Right Opening Door
Frame Color:
Anthracite
Blue
Red
Unit Size:
Each
Product Code. 30057587
Brand: BICASA KDORHB
Only null left
Estimated Shipment: 15-08-2025
Add to basket
Only null left
Add to basket
Suspended Undercounter cabinet with Hinged Door for Kompatto workstation. Made in Fire-retardant particleboard.
  • For Kompatto Series Workstation
  • Suspended under the countertop
  • Doors: 270° degrees opening - Automatic hold-closing hinges
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
  • Steel sheet shelf (height-adjustable) - Keyable lock as option
  • Fire-retardant particleboard body
  • Light Gray color (2 decorative) - mm 19 thickness
  • Frontal drawer panels and doors with ABS rounded rim
Specifications
Product Type Suspended Undercounter Cabinet
Color Light Gray
Frame Color Blue
Material Particleboard
No. of Shelves 1
Includes Right Opening Door
For Use With (Equipment) Kompatto Series Workstation
Dimensions (L x W x H) 600 x (500 + 20) x 530 mm
Width (English) 19.7 in.
Width (Metric) 500 mm
Product Content Correction
Product Title
BICASA Suspended Undercounter Cabinet with Hinged Door for Kompatto Workstation >