Learn More
SVL™ Glass Screwthreads
For use by glassblowers to construct apparatus with SVR™ joints
Brand:  SVL 703-03
 View more versions of this product
Product Code. 15517750
  • 4350.00 DKK / Pack of 50
Estimated Shipment: 07-10-2025
Explore more special offers
Description
  • For use by Glassblowers to construct apparatus with SVL™ joints
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3)
6.49 in.
2 mm
Junction Tube
32 mm
165 mm
For use by Glassblowers to construct apparatus with SVL™ joints
Product Title
SVL™ Glass Screwthreads > Thickness: 2.0mm; Outside dia.: 32mm; Joint Size:30