Learn More
BINDER™ Swivel Casters
Height-adjustable swivel casters
Brand:  BINDER™ 8012-2542
 View more versions of this product
Product Code. 30238323
  • 4803.14 DKK / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Description
Height-adjustable swivel casters for horizontal alignment of unit on uneven floors.
Specifications
Specifications
KB PRO 260, KB PRO 470, KB PRO 720, KB PRO 1060, KB PRO 1600
Steel
Castor Frame
Product Title
BINDER™ Swivel Casters > For use with KB PRO 260, KB PRO 470, KB PRO 720, KB PRO 1060, KB PRO 1600