missing translation for 'onlineSavingsMsg'
Learn More

KARTELL Sylicon Conical Stopper

The Sylicon Conical Stopper creates a seal in bottles and flasks.

Brand:  KARTELL 0381300

 View more versions of this product

Product Code. 10747672

  • 1085.00 DKK / Pack of 5
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Autoclavable.
  • SUITABLE FOR FOODSTUFF.
  • MATERIAL: SYLICON
Specifications

Specifications

Autoclavable
34 mm
45 mm
Non-sterile
White
45 mm
Silicone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers