Learn More
Hamilton™ Syringes
Brand: Hamilton™ 203330
This item is not returnable.
View return policy
Specifications
Not Autoclavable | |
1001 | |
1 mL |
Syringe | |
1/Pk. |
Product Content Correction
Product Title
Hamilton™ Syringes > Volume: 1mL
Spot an opportunity for improvement?Share a Content Correction