Learn More
Abnova™ SYTL3 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Brand:  Abnova™ H00094120-T01.100uL
Product Code. 16166883
  • 1710.00 DKK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
293T
SYTL3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 68.6kDa
Transient overexpression cell lysate was tested with anti-SYTL3 antibody (Mfr. No. H00094120-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
BC100830
pCMV-SYTL3 full-length
100 μL
Human