Learn More
BRAND™ T-Bore Stopcock
T-Bore Stopcock is constructed of PTFE and includes a nozzle.
471.00 DKK - 523.00 DKK
Specifications
Type T-Bore Stopcock
Material PTFE
Includes Nozzle
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10203781
View Documents
BRAND™
88660
3 mm
471.00 DKK
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
10691372
View Documents
BRAND™
88675
4 mm
523.00 DKK
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Polypropylene valve lever
  • Outstanding chemical resistance
  • With integrated hose nozzles
Specifications
Specifications
T-Bore Stopcock
PTFE
Nozzle
Videos
SDS
Documents
Documents
Product Certifications