Learn More
BRAND™ T-Shape Tubing Connector
Manufactured from translucent polypropylene, Brand™ T-Shape Tubing Connectors can be repeatedly autoclaved at 121°C.
Brand: BRAND™ 152815
This item is not returnable.
View return policy
Description
- Good chemical resistance and mechanical properties
- Straight-through tube and side arm
- Provided in packs of 20 (152815) and 10 (152825)
Specifications
Length of straight-through tube: 69mm; length of side arm including diameter of straight-through tube: 47mm | |
Polypropylene | |
20/Pk. |
Autoclavable | |
Tubing with inner diameter of 10-11mm | |
Translucent |