Learn More
LNI™ Tablo S 92 D Bench Product Code.: 17830937
Only undefined left
Add to basket
LNI™ Tablo S 92 D Bench
Product Code. 17830937 Shop All LNI Swissgas Products
Tablo S 92 D Bench; Dimensions (D x W x H): 890 x 920 x 840 mm
Click to view available options
:
Tablo S 92 D Bench; Dimensions (D x W x H): 890 x 920 x 840 mm
Product Code. 17830937
Brand: LNI™ 2000.02.011D
Only undefined left
Add to basket
Only undefined left
Add to basket
Instrument laboratory bench for LC, GC or ICP MS systems and LNI™ gas generators with chemical resistant top.
  • Designed without rouging pumps or noisy equipment
  • Made with chemical resistant top, side panels, castors for easy moving of the system, and adjustable leveling feet for rock solid stability of the bench
  • With space of various dimensions under the top, TABLO benches are the ideal solution to create a cleaner, tidier and more efficient lab workspace and save space
  • Space under top dimensions (D x W x H): 640 x 500 x 745 to 775 mm
Specifications
Product Type Bench
Color Light Gray
Adjustable Yes
Autoclavable Not Autoclavable
Dimensions (D x W x H) 890 x 920 x 840 mm
For Use With (Equipment) Analytical Instruments
Frame Material Stainless Steel
Includes Chemical Resistant Table Top
No. of Doors None
No. of Drawers None
No. of Shelves None
Plug Type EU-style, UK-style
Warranty 24 Months
With Casters Yes
Footrest No
With Tilt No
Material Stainless Steel
Show More Show Less
Product Content Correction
Product Title
LNI™ Tablo S 92 D Bench >