Learn More
Sartorius Tacta™ Mechanical Pipettes
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
Brand: Sartorius LH-729070
2121.30 DKK valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Description
- Comfortable to hold due to the ergonomically designed handle
- Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
- Controlled and smooth tip ejection with the new Sartorius Optiject technology
- Calibration adjustment to provide accurate results for various liquid types
- Easy to clean, with only three parts to disassemble
- Reliable results, even over lengthy pipetting periods
Specifications
Optifit 1000 μL, SafetySpace Filter Tips 1000 μL | |
Autoclavable | |
Reusable | |
Polypropylene | |
Tacta | |
Non-sterile | |
100 to 1000 μL |
Yes | |
Blue | |
Ergonomic | |
1 | |
No | |
Pipette | |
Yellow |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%