missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1706.22 DKK - 3498.83 DKK
Specifications
Antigen | TAF12 |
---|---|
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18306116
|
Bio-Techne
NBP3-03296-20UL |
20 μL |
1706.22 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310497
|
Bio-Techne
NBP3-03296-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAF12 Polyclonal specifically detects TAF12 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
TAF12 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
6883 | |
IgG | |
Affinity purified |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
20kDa, TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF15, TAF2J, TAFII-20/TAFII-15, TAFII20TAFII20/TAFII15, TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD, Transcription initiation factor TFIID 20/15 kDa subunits, transcription initiation factor TFIID subunit 12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human TAF12 (NP_001128690.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |