missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAZ Mouse anti-Human, Mouse, Rat, Clone: 2A12A10, Proteintech
Mouse Monoclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | TAZ |
---|---|
Clone | 2A12A10 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16848435
|
Proteintech
66500-1-IG-20UL |
20 ÎĽL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16838435
|
Proteintech
66500-1-IG-150UL |
150 ÎĽL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WW domain-containing transcription regulator protein 1 (WWTR1) is a transcriptional coactivator which acts as a downstream regulatory target in the Hippo signaling pathway that plays a pivotal role in organ size control and tumor supression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1. In turn, MOB1 phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. WWTR1 enhances PAX8 and NKX2-1/TTF1-dependent gene activation. WWTR1 regulates the nuclear accumulation of SMADS and has a key role in coupling them to the transcriptional machinery such as the mediator complex. WWTR1 also regulates embryonic stem-cell self-renewal and promotes cell proliferation and epithelial-mesenchymal transition.Specifications
TAZ | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
TAZ, WWTR1 | |
Wwtr1 | |
IgG1 | |
Protein G | |
Antibody |
2A12A10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q9EPK5, Q9GZV5 | |
25937, 295062, 97064 | |
TAZ Fusion Protein Ag13330 | |
Primary | |
-20°C | |
WWTR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TAZ Mouse anti-Human, Mouse, Rat, Clone: 2A12A10, Proteintech