missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBE 10X Buffer, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
1 L
Description
- TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis, after dilution to working concentrations.
- TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.
- TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.
- TBE buffer maintains the structural integrity of nucleic acids and more suitable for their size analysis.
- TBE has a greater buffering capacity and will give sharper resolution. TBE is better suited for high-voltage (>150 V) electrophoresis because of its higher buffering capacity.

Specifications
Specifications
Color | Clear Colorless |
pH | 8.2 to 8.4 (literature) |
Physical Form | Solution |
Quantity | 1 L |
Solubility Information | Soluble in water. |
Safety and Handling

Product Identifier
- TBE 10X BUFFER
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Boric Acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TBE 10X Buffer, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction