missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC-MCH 7c, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Potent, selective melanin-concentrating hormone receptor 1 (MCH1R) antagonist (IC50= 5.6 nM in hMCH1R-expressing CHO cells). Displays selectivity for MCH1R over MCH2R (IC50 = > 10 μM). Decreases body weight in a mouse model of diet-induced obesity. Brain penetrant; orally active.
Specifications
Specifications
CAS | 864756-35-4 |
Molecular Formula | C24H25FN2O3 |
Quantity | 10 mg |
Synonym | tc-mch 7c, 4-4-fluorobenzyloxy-1-4-2-pyrrolidin-1-yl ethoxy phenyl pyridin-2 1h-one, 4-4-fluorophenyl methoxy-1-4-2-1-pyrrolidinyl ethoxy phenyl-2 1h-pyridinone |
InChI Key | ANCFKYJMXNMYNZ-UHFFFAOYSA-N |
SMILES | C1CCN(C1)CCOC2=CC=C(C=C2)N3C=CC(=CC3=O)OCC4=CC=C(C=C4)F |
IUPAC Name | 4-[(4-fluorophenyl)methoxy]-1-[4-(2-pyrrolidin-1-ylethoxy)phenyl]pyridin-2-one |
Molecular Weight (g/mol) | 408.473 |
PubChem CID | 11654412 |
Formula Weight | 408.47 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'