Learn More
Teknorinse™ Acidid Rinse Aid
Teknorinse enables water to drain evenly from washed items so that they are left virtually dry
Brand:  Teknon TK310/5
Product Code. 12701895
  • 636.00 DKK / 5 L
Explore more special offers
This item is not returnable. View return policy
Description
Description
Teknorinse enables water to drain evenly from washed items so that they are left virtually dry. It also helps to neutralise the alkaline detergent used for the wash cycle. This formulation is designed to operate in hard or soft water and helps prevent scale and promotes efficiency. It is highly economical having an in-use concentration in the order of 300ppm.
  • Rinse neutraliser
  • Scale control
  • Low foam
  • Free rinsing An acidic rinse aid for use in laboratory, medical and industrial automatic washing machines.
Specifications
Specifications
Teknorinse Acidic Rinse Aid
For use in laboratory, medical and industrial automatic washing machines
5 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Teknorinse™ Acidid Rinse Aid > Quantity: 5L