Learn More
BINDER™ Temperature and Humidity Calibration Certificate
Calibration certificate including 27 temperature measurements and 1 humidity measurement.
20801.17 DKK
Specifications
Certifications/Compliance | DIN12880 |
---|---|
Duration | 1 Day |
Product Type | Calibration Certificate |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
30238362
|
BINDER™
8012-2577 |
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600 |
20801.17 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30238306
|
BINDER™
8012-2578 |
KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 |
20801.17 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- 27 temperature measuring points
- 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
DIN12880 | |
1 Day | |
Calibration Certificate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BINDER™ Temperature and Humidity Calibration Certificate