missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Tenofovir Product Code.: 16607042
compliance-icons

Thermo Scientific Chemicals Tenofovir

Product Code. 16607042
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 16607042

Brand: Thermo Scientific Chemicals 465610050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147127-20-6
Molecular Formula C9 H14 N5 O4 P

Specifications

Quantity 5 g
Formula Weight 287.21
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key SGOIRFVFHAKUTI-ZCFIWIBFSA-N
SMILES C[C@H](CN1C=NC2=C(N)N=CN=C12)OCP(O)(O)=O
IUPAC Name ({[(2R)-1-(6-amino-9H-purin-9-yl)propan-2-yl]oxy}methyl)phosphonic acid
Molecular Weight (g/mol) 287.22
Chemical Name or Material Tenofovir

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468932 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number465610010,465610050
Lot NumberA0460898 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number465610010,465610050
Lot NumberA0435209 Certificate TypeCertificate of Analysis Date11/10/2021 Catalog Number465610010,465610050
Lot NumberA0416618 Certificate TypeCertificate of Analysis Date30/08/2021 Catalog Number
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Tenofovir >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.