missing translation for 'onlineSavingsMsg'
Learn More
TEP 10 Model Buffer Solution pH 10, WTW™ Product Code.: 10312721

TEP 10 Model Buffer Solution pH 10, WTW™

Product Code. 10312721
1000 mL, 1 L
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 10312721

Brand: WTW 108704

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

pH buffers by WTW are precise and traceable to PTB/NIST in bulk size of 1 liter with built-in dosing vessel standard buffer with an accuracy that is ± 0,03 pH.

  • TEP 10 Model Buffer Solution pH 10
  • pH buffers by WTW are precise and traceable to PTB/NIST in bulk size of 1 liter with built-in dosing vessel Standard buffer
  • Live up to 3 years (unopened)
  • Thrifty use
  • Easiest handling by means of dosing device
TRUSTED_SUSTAINABILITY

Specifications

pH 10
Physical Form Liquid
Quantity 1000 mL
Packaging Bottle
Traceability to NIST Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TEP 10 Model Buffer Solution pH 10, WTW™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.