missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 97% Product Code.: 10001664

tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 97%

Product Code. 10001664
Amber Glass Bottle; 5g
Click to view available options
Amber Glass Bottle; 5g
This item is not returnable. View return policy

Product Code. 10001664

Brand: Thermo Scientific Maybridge AW00470DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 77279-24-4
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H22N2O3
Quantity 5 g
Synonym tert-butyl 4-2-hydroxyethyl piperazine-1-carboxylate, 1-boc-4-2-hydroxyethyl piperazine, 1-tert-butyloxy carbonyl-4-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-hydroxyethyl-piperazine, 4-2-hydroxy-ethyl-piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-2-hydroxyethyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-2-hydroxyethyl piperazine, tert-butyl 4-2-hydroxyethyl-1-piperazinecarboxylate, 4-2-hydroxyethyl piperazine-1-carboxylic acid tert-butyl ester
InChI Key VRXIOAYUQIITBU-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CCO
IUPAC Name tert-butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
Molecular Weight (g/mol) 230.308
PubChem CID 735736
Formula Weight 230.31
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.